missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Ribonuclease A His Protein

Product Code. 18348412 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
100 μg
20 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
20µg
250µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18348412 20 μg 20µg
18399102 50 μg 50µg
18374922 500 μg 500µg
18367982 250 μg 250µg
18389662 10 μg 10µg
18390702 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 18348412 Supplier Novus Biologicals™ Supplier No. NBP31216420UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specific activity is > 3 X 10^6 unit/mg, and is defined as the amount of enzyme that cleaves 1.0 nmole of RNA per minute at 25C.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/ml
For Use With (Application) Bioactivity, SDS-PAGE
Formulation PBS (pH 7.4), 10% glycerol
Gene ID (Entrez) 6035
Molecular Weight (g/mol) M.W. Theoretical: 15.3 kDa
Quantity 20 μg
Storage Requirements Store at 4C short term. Aliquot and store at -70C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration <1 EU per 1ug of protein (determined by LAL method)
Gene Alias EC 3.1.27.5, HP-RNase, MGC12408, RIB1, RIB-1, Ribonuclease 1, Ribonuclease A, ribonuclease pancreatic, ribonuclease, RNase A family, 1 (pancreatic), RNase A, RNase UpI-1, RNS1RNase 1
Gene Symbol RNASE1
Research Category DNA replication Transcription Translation and Splicing, Proteases & Other Enzymes
Purity or Quality Grade >90%, by SDS-PAGE
Protein Ribonuclease A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.