missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Resistin / ADSF / RETN Protein

Product Code. 15945759
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
5 μg
500 μg
Unit Size:
100µg
1mg
25µg
500µg
5µg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15945759 5 μg 5µg
15925759 25 μg 25µg
15905759 100 μg 100µg
15935759 500 μg 500µg
15915759 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15945759 Supplier enQuireBio™ Supplier No. QP52365ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number NP_001180303.1
Gene ID (Entrez) 56729
Name Resistin / ADSF / RETN Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol RETN
Biological Activity Human Resistin / ADSF / RETN induces ERK phosphorylation and activity (pERK) in SH-SY5Y cells via western blot (significanty different from control at Human Resistin / ADSF / RETN concentrations greater than 100 ng/ml). Activity was further confirmed by measuring IL-8 production in human PBMCs (typical ED50 is less than 500 ng/ml).
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence MSSKTLCSME EAINERIQEV AGSLIFRAIS SIGLECQSVT SRGDLATCPR GFAVTGCTCG SACGSWDVRA ETTCHCQCAG MDWTGARCCR VQP
Buffer Sterile filtered (0.22 um) solution containing 0.1% TFA
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.