missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Resistin / ADSF / RETN Protein
Click to view available options
Quantity:
1 mg
100 μg
25 μg
5 μg
500 μg
Dimensione della confezione:
100µg
1mg
25µg
500µg
5µg
Specifica
Specifica
| Accession Number | NP_001180303.1 |
| Gene ID (Entrez) | 56729 |
| Name | Resistin / ADSF / RETN Protein |
| Quantity | 1 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
| Gene Symbol | RETN |
| Biological Activity | Human Resistin / ADSF / RETN induces ERK phosphorylation and activity (pERK) in SH-SY5Y cells via western blot (significanty different from control at Human Resistin / ADSF / RETN concentrations greater than 100 ng/ml). Activity was further confirmed by measuring IL-8 production in human PBMCs (typical ED50 is less than 500 ng/ml). |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Vedi altri risultati |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto