missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Renalase Protein

Product Code. 18390358
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18390358 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18390358 Supplier R&D Systems™ Supplier No. 9887RE050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Renalase Protein is derived from E. coli. The Recombinant Human Renalase Protein has been validated for the following applications: Enzyme Activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q5VYX0-1
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2μm filtered solution in Tris, EDTA, DTT and Glycerol.
Gene ID (Entrez) 55328
Molecular Weight (g/mol) 39 kDa
Name C10orf59, C10orf59chromosome 10 open reading frame 59, EC 1.4, FLJ11218, MAO-C, Monoamine oxidase-C, RENALASE, renalase, FAD-dependent amine oxidase, RNLS
Quantity 50 μg
Source E. coli-derived human Renalase protein Ala2-Ile342 with substitution Glu37Asp,N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Species Human
Activity Measured by its ability to reduce Resazurin. The specific activity is >3.5 pmol/min/μg, as measured under the described conditions.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.