missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Relaxin-3 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
£211.66
Specifications
Gene ID (Entrez) | 117579 |
---|---|
Molecular Weight (g/mol) | 2.46 kDa (A chain), 3.04 kDa (B chain) |
Name | Relaxin-3 |
Quantity | 25μg |
Source | E. coli-derived human Relaxin-3 protein Asp119-Cys142 (A chain) & Ala27-Arg53 (B chain) |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18108190
|
R&D Systems™
3107-RN-025 |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
£211.66
25µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18136770
|
R&D Systems™
3107-RN-025/CF |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. |
£211.66
25µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Human Relaxin-3 Protein is derived from E. coli. The Recombinant Human Relaxin-3 Protein has been validated for the following applications: Bioactivity.Specifications
117579 | |
Relaxin-3 | |
E. coli-derived human Relaxin-3 protein Asp119-Cys142 (A chain) & Ala27-Arg53 (B chain) | |
Disulfide-linked heterodimer | |
Recombinant |
2.46 kDa (A chain), 3.04 kDa (B chain) | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |