missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Regulator of G-protein signaling 10 Protein
A DNA sequence encoding the Homo sapiens (Human) Regulator of G-protein signaling 10, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6602-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001005339.1 | |
Regulator of G-protein signaling 10 Protein | |
Research Use Only | |
RGS10 | |
Recombinant Protein | |
E. coli | |
MFNRAVSRLS RKRPPSDIHD SDGSSSSSHQ SLKSTAKWAA SLENLLEDPE GVKRFREFLK KEFSEENVLF WLACEDFKKM QDKTQMQEKA KEIYMTFLSS KASSQVNVEG QSRLNEKILE EPHPLMFQKL QDQIFNLMKY DSYSRFLKSD LFLKHKRTEE EEEDLPDAQT AAKRASRIYN T | |
Greater than 80% as determined by SDS-PAGE. |
6001 | |
50 μg | |
Not determined. | |
Full Length of Isoform 3 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |