missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Reelin Protein

Product Code. 18736157
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18736157

Brand: R&D Systems™ 8546MR050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Reelin Protein is derived from CHO. The Recombinant Human Reelin Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P78509
For Use With (Application) Bioactivity
Gene ID (Entrez) 5649
Molecular Weight (g/mol) Observed M.W.: 155-188kDa, reducing conditions, Theoretical M.W.: 163kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived human Reelin protein Ser1221-Gln2666,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias EC 3.4.21, EC 3.4.21.-, Reeler, reelin, RELN, RL, RLPRO1598
Common Name Reelin
Gene Symbol RELN
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Human Reelin is coated at 1.5μg/mL (100μL/well), the concentration of Recombinant Human Apolipoprotein E R2/ApoE R2 that produces 50% optimal binding response is typically 50-300ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Ser1221-Gln2666, with a C-terminal 6-His tag
Purity or Quality Grade >90%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.