missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Reelin Protein

A DNA sequence encoding the Homo sapiens (Human) Reelin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6597-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16064430

  • £384.00 / 100µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_005036.2
Reelin Protein
Research Use Only
RELN
Recombinant Protein
E. coli
AAGYYPRFSP FFFLCTHHGE LEGDGEQGEV LISLHIAGNP TYYVPGQEYH VTISTSTFFD GLLVTGLYTS TSVQASQSIG GSSAFGFGIM SDHQFGNQFM CSVVASHVSH LPTTNLSFIW IAPPAGTGCV NFMATATHRG QVIFKDALAQ QLCEQGAPTD VTVHPHLAEI HSDSIILRDD FDSYHQLQLN PNIWVECNNC ETGEQCGAIM HGNAVTFCEP YGPRELITT
Greater than 90% as determined by SDS-PAGE.
5649
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers