missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RCHY1 Protein

Product Code. 16001151
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16001151 50 μg 50µg
16061141 10 μg 10µg
16051141 100 μg 100µg
16081141 200 μg 200µg
16091141 500 μg 500µg
16071141 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16001151 Supplier enQuireBio™ Supplier No. QP7847ec50ug
Quantity Price
1 £214.03 / 50µg
6 £198.60 / 50µg

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) RCHY1, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001009922.1
Gene ID (Entrez) 25898
Name RCHY1 Protein
Quantity 50 μg
Regulatory Status RUO
Endotoxin Concentration Not determined.
Gene Symbol RCHY1
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag GST
Sequence MAATAREDGA SGQERGQRGC EHYDRGCLLK APCCDKLYTC RLCHDNNEDH QLDRFKVKEV QCINCEKIQH AQQTCEECST LFGEYYCDIC HLFDKDKKQY HCENCGICRI GPKEDFFHCL KCNLCLAMNL QGRHKCIENV SRQNCPICLE DIHTSRVVAH VLPCGHLLHR TCYEEMLKEG YRCPLCMHSA LDMTRYWRQL DDEVAQTPMP SEYQNMTVDI LCNDCNGRST VQFHILGMKC KICESYNTAQ AGGRRISLDQ Q
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.