missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human RCBTB2 GST (N-Term) Protein

Product Code. 18333928 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18333928 10 μg 10µg
18380828 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18333928 Supplier Novus Biologicals™ Supplier No. H00001102P0110ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formulation 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 1102
Molecular Weight (g/mol) M.W. (Theoretical): 86.7 kDa
Quantity 10 μg
Storage Requirements Store at -80C. Avoid freeze-thaw cycles.
Gene Alias CHC1-L, CHC1LRCC1-like G exchanging factor RLG, Chromosome condensation 1-like, RCC1 and BTB domain-containing protein 2, RCC1-like G exchanging factor, regulator of chromosome condensation (RCC1) and BTB (POZ) domain containingprotein 2, regulator of chromosome condensation and BTB domain containing protein 2, Regulator of chromosome condensation and BTB domain-containing protein 2, RLG
Gene Symbol RCBTB2
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein RCBTB2
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.