missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ras-related protein R-Ras2 Protein
A DNA sequence encoding the Homo sapiens (Human) Ras-related protein R-Ras2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6628-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001096139.1 | |
Ras-related protein R-Ras2 Protein | |
Research Use Only | |
RRAS2 | |
Recombinant Protein | |
E. coli | |
AAAGWRDGSG QEKYRLVVVG GGGVGKSALT IQFIQSYFVT DYDPTIEDSY TKQCVIDDRA ARLDILDTAG QEEFGAMREQ YMRTGEGFLL VFSVTDRGSF EEIYKFQRQI LRVKDRDEFP MILIGNKADL DHQRQVTQEE GQQLARQLKV TYMEASAKIR MNVDQAFHEL VRVIRKFQEQ ECPPSPEPTR KEKDKKGCHC | |
Greater than 80% as determined by SDS-PAGE. |
22800 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |