missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ras-related protein R-Ras2 Protein

A DNA sequence encoding the Homo sapiens (Human) Ras-related protein R-Ras2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6628-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16004660

  • £1735.00 / 1mg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001096139.1
Ras-related protein R-Ras2 Protein
Research Use Only
RRAS2
Recombinant Protein
E. coli
AAAGWRDGSG QEKYRLVVVG GGGVGKSALT IQFIQSYFVT DYDPTIEDSY TKQCVIDDRA ARLDILDTAG QEEFGAMREQ YMRTGEGFLL VFSVTDRGSF EEIYKFQRQI LRVKDRDEFP MILIGNKADL DHQRQVTQEE GQQLARQLKV TYMEASAKIR MNVDQAFHEL VRVIRKFQEQ ECPPSPEPTR KEKDKKGCHC
Greater than 80% as determined by SDS-PAGE.
22800
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers