missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human RAP30 His Protein

Product Code. 18328634 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mg
100 μg
Unit Size:
0.50mg
100µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18328634 100 μg 100µg
18382992 0.5 mg 0.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18328634 Supplier Novus Biologicals™ Supplier No. NBP317983100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5 mg/ml
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.2 M NaCl, 1 mM DTT
Gene ID (Entrez) 2963
Molecular Weight (g/mol) M.W. Theoretical: 30.5 kDa
Quantity 100 μg
Storage Requirements Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles.
Gene Alias ATP-dependent helicase GTF2F2, BTF4, General transcription factor IIF 30 kDa subunit, general transcription factor IIF subunit 2, general transcription factor IIF, polypeptide 2, 30kDa, TF2F2, TFIIF, TFIIF-beta, Transcription initiation factor IIF subunit beta, Transcription initiation factor RAP30
Gene Symbol GTF2F2
Research Category Chromatin Research
Purity or Quality Grade >90%, by SDS-PAGE
Protein RAP30
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.