missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RAP2A Protein

A cDNA sequence encoding the RAP2A was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13267-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15943029

  • £5730.00 / 1mg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

RAP2A Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MREYKVVVLG SGGVGKSALT VQFVTGTFIE KYDPTIEDFY RKEIEVDSSP SVLEILDTAG TEQFASMRDL YIKNGQGFIL VYSLVNQQSF QDIKPMRDQI IRVKRYEKVP VILVGNKVDL ESEREVSSSE GRALAEEWGC PFMETSAKSK TMVDELFAEI VRQMNYAAQP DKDDPCCSAC
Greater than 95.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The RAP2A solution (1 mg/ml) 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers