Learn More
enQuireBio™ Recombinant Human RANKL / OPGL / TNFSF11 / CD254 Protein
Recombinant Protein
Brand:  enQuireBio™ QP5325-100ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15945909
  • £775.22 / 100µg
Estimated Shipment: 01-03-2022
Specifications
NP_003692.1
RANKL / OPGL / TNFSF11 / CD254 Protein
100 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Recombinant Protein
E. coli
EKAMVDGSWL DLAKRSKLEA QPFAHLTINA TDIPSGSHKV SLSSWYHDRG WAKISNMTFS NGKLIVNQDG FYYLYANICF RHHETSGDLA TEYLQLMVYV TKTSIKIPSS HTLMKGGSTK YWSGNSEFHF YSINVGGFFK LRSGEEISIE VSNPSLLDPD QDATYFGAFK VRDID
8600
Greater than 95% as determined by SDS-PAGE.
Research Use Only
TNFSF11
Human
Untagged
10 mM sodium phosphate, pH 7.5