missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RAMP3 Protein

A cDNA sequence encoding the RAMP3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13260-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15943009

  • £99.15 / 2µg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

10268
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MRAGGCNETG MLERLPLCGK AFADMMGKVD VWKWCNLSEF IVYYESFTNC TEMEANVVGC YWPNPLAQGF ITGIHRQFFS NCTVDRVHLE DPPDEV
Greater than 90% as determined by SDS-PAGE.
RAMP3 Protein
Research Use Only
RAMP3
Human
His
RAMP3 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.1M NaCl.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers