missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RALY Protein
A cDNA sequence encoding the RALY was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13258-5ug
Additional Details : Weight : 0.01000kg
Specifications
RALY Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSLKLQA SNVTNKNDPK SINSRVFIGN LNTALVKKSD VETIFSKYGR VAGCSVHKGY AFVQYSNERH ARAAVLGENG RVLAGQTLDI NMAGEPKPDR PKGLKRAASA IYSGYIFDYD YYRDDFYDRL FDYRGRLSPV PVPRAVPVKR PRVTVPLVRR VKTNVPVKLF ARSTAVTTSS AKIKLKSSEL QAIKTELTQI KSNIDALLSR LEQIAAEQKA NPDGKKKGDG GGAGGGGGGG GSGGGGSGGG GGGGSSRPPA PQENTTSEAG LPQGEARTRD DGDEEGLLTH SEEELEHSQD TDADDGALQ | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RALY protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |