missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Rab6 / Rab GTPases / RAB6A Protein
A DNA sequence encoding the Homo sapiens (Human) Rab6 / Rab GTPases / RAB6A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6574-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001230647.1 | |
Rab6 / Rab GTPases / RAB6A Protein | |
Research Use Only | |
RAB6A | |
Recombinant Protein | |
E. coli | |
STGGDFGNPL RKFKLVFLGE QSVGKTSLIT RFMYDSFDNT YQATIGIDFL SKTMYLEDRT VRLQLWDTAG QERFRSLIPS YIRDSTVAVV VYDITNVNSF QQTTKWIDDV RTERGSDVII MLVGNKTDLA DKRQVSIEEG ERKAKELNVM FIETSAKAGY NVKQLFRRVA AALPGMESTQ DRSREDMIDI KLEKPQEQPV SEGGCSC | |
Greater than 90% as determined by SDS-PAGE. |
5870 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |