missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Rab5a Protein
Shop All Bio Techne Products
Click to view available options
:
0.1mg; Unlabeled
Description
An un-tagged recombinant protein corresponding to the amino acids 1-215 of Human Rab5a The Recombinant Human Rab5a Protein is derived from E. coli. The Recombinant Human Rab5a Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Accession Number | NP_004153 |
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | 20mM Tris buffer (pH 8.0) |
| Gene ID (Entrez) | 5868 |
| Molecular Weight (g/mol) | 23kDa |
| Name | Rab5a Protein |
| Purification Method | Protein |
| Immunogen | MASRGATRPNGPNTGNKICQFKLVLLGESAVGKSSLVLRFVKGQFHEFQESTIGAAFLTQTVCLDDTTVKFEIWDTAGQERYHSLAPMYYRGAQAAIVVYDITNEESFARAKNWVKELQRQASPNIVIALSGNKADLANKRAVDFQEAQSYADDNSLLFMETSAKTSMNVNEIFMAIAKKLPKNEPQNPGANSARGRGVDLTEPTQPTRNQCCSN |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction