missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Rab5 GDP / GTP exchange factor Protein
A DNA sequence encoding the Homo sapiens (Human) Rab5 GDP/GTP exchange factor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8190-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001273989.1 | |
Rab5 GDP / GTP exchange factor Protein | |
Research Use Only | |
RABGEF1 | |
Recombinant Protein | |
E. coli | |
MSLKSERRGI HVDQSDLLCK KGCGYYGNPA WQGFCSKCWR EEYHKARQKQ IQEDWELAER LQREEEEAFA SSQSSQGAQS LTFSKFEEKK TNEKTRKVTT VKKFFSASSR VGSKKEIQEA KAPSPSINRQ TSIETDRVSK EFIEFLKTFH KTGQEIYKQT KLFLEGMHYK RDLSIEEQSE CAQDFYHNVA ERMQTRGKVP PERVEKIMDQ IEKYIMTRLY KYVFCPETTD DEKKDLAIQK RIRALRWVTP QMLCVPVNED IPEVSDMVVK AITDIIEMDS KRVPRDKLAC ITKCSKHIFN AIKITKNEPA SADDFLPTLI YIVLKGNPPR LQSNIQYITR FCNPSRLMTG EDGYYFTNLC CAVAFIEKLD AQSLNLSQED FDRYMSGQTS PRKQEAESWS PDACLGVKQM YKNLDLLSQL NERQERIMNE AKKLEKDLID WTDGIAREVQ DIVEKYPLEI KPPNQPLAAI DSENVENDKL PPPLQPQVYA G | |
Greater than 90% as determined by SDS-PAGE. |
27342 | |
500 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |