missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAB2A Protein
A DNA sequence encoding the Homo sapiens (Human) RAB2A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6569-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001229573.1 | |
RAB2A Protein | |
Research Use Only | |
RAB2A | |
Recombinant Protein | |
E. coli | |
AYAYLFKYII IGDTGVGKSC LLLQFTDKRF QPVHDLTIGV EFGARMITID GKQIKLQIWD TAGQESFRSI TRSYYRGAAG ALLVYDITRR DTFNHLTTWL EDARQHSNSN MVIMLIGNKS DLESRREVKK EEGEAFAREH GLIFMETSAK TASNVEEAFI NTAKEIYEKI QEGVFDINNE ANGIKIGPQH AATNATHAGN QGGQQAGGGC C | |
Greater than 90% as determined by SDS-PAGE. |
5862 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |