missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAB21 Protein
A cDNA sequence encoding the RAB21 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13232-10ug
Additional Details : Weight : 0.01000kg
Specifications
RAB21 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMYSF KVVLLGEGCV GKTSLVLRYC ENKFNDKHIT TLQASFLTKK LNIGGKRVNL AIWDTAGQER FHALGPIYYR DSNGAILVYD ITDEDSFQKV KNWVKELRKM LGNEICLCIV GNKIDLEKER HVSIQEAESY AESVGAKHYH TSAKQNKGIE ELFLDLCKRM IETAQVDERA KGNGSSQPGT ARRGVQIIDD EPQAQTSGGG CC | |
Greater than 90.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RAB21 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 2mM DTT, 200mM NaCl and 1mM PMSF. |