missing translation for 'onlineSavingsMsg'
Learn More

R&DSystems™ Recombinant Human R-Spondin 3 Fc Chimera Protein, CF

Product Code. 30023122
Change view
Click to view available options
Quantity:
100 μg
20 μg
50 μg
500 μg
Unit Size:
100µg
20µg
500µg
50µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30023122 500 μg 500µg
30023117 100 μg 100µg
30023123 50 μg 50µg
30023140 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30023122 Supplier R&DSystems™ Supplier No. 11506RS500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Measured by its ability to activate TCF reporter activity in HEK293 human embryonic kidney cells in the presence of Recombinant Human Wnt-3a (Catalog # 5036-WN ). The ED50 for this effect is 0.500-10.0 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BXY4.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 84870
Molecular Weight (g/mol) M.W.-observed: 53-69 kDa, under reducing conditions., M.W.-theroretical: 46 kDa
Quantity 500 μg
Source Chinese Hamster Ovary cell line, CHO-derived human R-Spondin 3 protein Human RSPO3 (Met33-Gly209) (N-terminus) Accession # Q9BXY4.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias Cristin 1, CRISTIN1, FLJ14440, hPWTSR, hRspo3, Protein with TSP type-1 repeat, PWTSR, Roof plate-specific spondin-3, RSPO3, RSpondin 3, R-spondin 3 homolog (Xenopus laevis), R-spondin-3, Thrombospondin type-1 domain-containing protein 2, thrombospondin, type I, domain containing 2, THSD2
Conjugate Unconjugated
Reconstitution Reconstitute the 20 μg size at 200 μg/mL in PBS. Reconstitute all other sizes at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein R-Spondin 3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.