missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human QTRTD1 Protein

Product Code. 15912909
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
This item is not returnable. View return policy

Product Code. 15912909

Brand: enQuireBio™ QP132271mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the QTRTD1 was constructed and used to recombinantly synthesize the protein.

Specifications

Name QTRTD1 Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MGSMKLSLTK VVNGCRLGKI KNLGKTGDHT MDIPGCLLYT KTGSAPHLTH HTLHNIHGVP AMAQLTLSSL AEHHEVLTEY KEGVGKFIGM PESLLYCSLH DPVSPCPAGY VTNKSVSVWS VAGRVEMTVS KFMAIQKALQ PDWFQCLSDG EVSCKEATSI KRVRKSVDRS LLFLDNCLRL QEESEVLQKS VIIGVIEGGD VMEERLRSAR ETAKRPVGGF LLDGFQGNPT TLEARLRLLS SVTAELPEDK PRLISGVSRP DEVLECIERG VDLFESFFPY QVTERGCALT FSFDYQPNPE ETLLQQNGTQ EEIKCMDQIK KIETTGCNQE ITSFEINLKE KKYQEDFNPL VRGCSCYCCK NHTRAYIHHL LVTNELLAGV LLMMHNFEHY FGFFHYIREA LKSDKLAQLK ELIHRQAS
Buffer QTRTD1 protein solution (0.25 mg/ml) containing Phosphate buffered saline (pH 7.4) 30% glycerol, 2mM DTT, 1mM EDTA and 0.1mM PMSF.
Purity or Quality Grade Greater than 95.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.