missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human QPCTL Protein

A DNA sequence encoding the Homo sapiens (Human) QPCTL, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8096-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16002651

  • £1735.00 / 1mg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001156849.1
QPCTL Protein
Research Use Only
QPCTL
Recombinant Protein
E. coli
AAPVTLQLLF LDGEEALKEW GPKDSLYGSR HLAQLMESIP HSPGPTRIQA IELFMLLDLL GAPNPTFYSH FPRTVRWFHR LRSIEKRLHR LNLLQSHPQE VMYFQPGEPF GSVEDDHIPF LRRGVPVLHL ISTPFPAVWH TPADTEVNLH PPTVHNLCRI LAVFLAEYLG L
Greater than 90% as determined by SDS-PAGE.
54814
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers