missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PYCR2 Protein

A cDNA sequence encoding the PYCR2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13221-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15932889

  • £155.00 / 20µg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

PYCR2 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSVGFIG AGQLAYALAR GFTAAGILSA HKIIASSPEM NLPTVSALRK MGVNLTRSNK ETVKHSDVLF LAVKPHIIPF ILDEIGADVQ ARHIVVSCAA GVTISSVEKK LMAFQPAPKV IRCMTNTPVV VQEGATVYAT GTHALVEDGQ LLEQLMSSVG FCTEVEEDLI DAVTGLSGSG PAYAFMALDA LADGGVKMGL PRRLAIQLGA QALLGAAKML LDSEQHPCQL KDNVCSPGGA TIHALHFLES GGFRSLLINA VEASCIRTRE LQSMADQEKI SPAALKKTLL DRVKLESPTV STLTPSSPGK LLTRSLALGG KKD
Greater than 90.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
PYCR2 protein solution (0.25 mg/ml) in phosphate buffered saline (pH 7.4), 50% glycerol, 5mM DTT and 1mM EDTA.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers