missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human PVRIG Fc Chimera Protein

Product Code. 18759057
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18759057 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18759057 Supplier R&D Systems™ Supplier No. 9365PV050
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Expressed in lymphocytes, such as T cells and NK cells, but not in monocyte derived dendritic cells.

Human PVRIG (poliovirus receptor related immunoglobulin domain-containing protein), also known as CD112 receptor (CD112R), is approximately 34 kDa single transmembrane protein in the poliovirus receptor-like protein (PVR) family. It is composed of single extracellular IgV domain, one transmembrane domain, and long intracellular domain. The intracellular domain contains two tyrosine residues, one within an ITIM-like motif that is potential docking site for phosphatases. Reconstitute at 500μg/mL in PBS

Storage:

Store the unopened product at −20 to −70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.

Order Info

Ships at ambient temperature.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6DKI7
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl and PEG with Trehalose.
Gene ID (Entrez) 79037
Molecular Weight (g/mol) 40 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human PVRIG protein, (N-terminus) Human PVRIG (Thr41 & Glu43-Leu172) Accession # Q6DKI7 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Biological Activity 0.4 to 2.4μg/mL
Product Type Recombinant protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 250 μg/mL in sterile water.
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.