missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Putative inactive neutral ceramidase B Protein

A DNA sequence encoding the Homo sapiens (Human) Putative inactive neutral ceramidase B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5682-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15946969

  • £384.00 / 100µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001072984.1
Putative inactive neutral ceramidase B Protein
Research Use Only
ASAH2B
Recombinant Protein
E. coli
MRQHRQFMDR THYLLTFSSS ETLLRLLLRI VDRAPKGRTF GDVLQPAKPE YRVGEVAEVI FVGANPKNSV QNQTHQTFLT VEKYEATSTS WQIVCNDASW ETRFYWHKGL LGLSNATVEW HIPDTAQPGI YRIRYFGHNR KQDILKPAVI LSFEGTSPAF EVVTI
Greater than 90% as determined by SDS-PAGE.
653308
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers