missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Specifications
Specifications
| For Use With (Application) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Formulation | 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Gene ID (Entrez) | 80324 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 69.63 kDa |
| Quantity | 25 μg |
| Storage Requirements | Store at -80°C. Avoid freeze/thaw cycles. |
| Gene Alias | EC 5.4.99.12, MGC11268, mitochondrial tRNA pseudouridine synthase A, MLASA1, pseudouridylate synthase 1, tRNA pseudouridine synthase A, mitochondrial, tRNA pseudouridylate synthase I, tRNA uridine isomerase I, tRNA-uridine isomerase I |
| Gene Symbol | PUS1 |
| Research Category | Core ESC Like Genes, Stem Cell Markers |
| Product Line | Novus |
| Show More |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?