missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human PTPN7 His Protein

Product Code. 18401071 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18401071 50 μg 50µg
18482301 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18401071 Supplier Novus Biologicals™ Supplier No. NBP19906850ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5 mg/ml
For Use With (Application) PAGE
Formulation 20 mM Tris-HCl buffer (pH8.0), 20% glycerol, 0.1 M NaCl, 1 mM DTT
Gene ID (Entrez) 5778
Molecular Weight (g/mol) 43.1 kDa
Quantity 50 μg
Storage Requirements Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Gene Alias BPTP-4, FLJ18703, HEPTPnonreceptor-type, stress induced, LPTP, protein tyrosine phosphatase, non-receptor type 7, tyrosine-protein phosphatase non-receptor type 7
Gene Symbol PTPN7
Research Category Cancer
Purity or Quality Grade >90% by SDS-PAGE
Protein PTPN7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.