missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PTP4A1 Protein

A cDNA sequence encoding the PTP4A1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13208-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15942839

  • £2975.00 / 1mg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

PTP4A1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MARMNRPAPV EVTYKNMRFL ITHNPTNATL NKFIEELKKY GVTTIVRVCE ATYDTTLVEK EGIHVLDWPF DDGAPPSNQI VDDWLSLVKI KFREEPGCCI AVHCVAGLGR APVLVALALI EGGMKYEDAV QFIRQKRRGA FNSKQLLYLE KYRPKMRLRF KDSNGHRNNC
Greater than 90.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The PTP4A1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers