missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human PTHLH/PTHrP 15N Stable Isotope Protein
Shop All Bio Techne ProductsSpecifications
Specifications
| Accession Number | P12272, P12272, P12272 |
| For Use With (Application) | PAGE |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 10 mM PB, pH 6.0, 300 mM NaCl. |
| Gene ID (Entrez) | 5744 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 10.0 kDa |
| Quantity | 1000 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/μg of PTHLH/PTHrP as determined by LAL method. |
| Gene Alias | BDE2, HHM, MGC14611, parathyroid hormone-like hormone, Parathyroid hormone-like protein, parathyroid hormone-related protein, PLPosteostatin, PTHR, PTH-related protein, PTHrP, PTH-rP, PTHRPparathyroid hormone-like related protein |
| Gene Symbol | PTHLH |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?