missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human PTH Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
1000 μg
500 μg
Unit Size:
1000µg
100µg
500µg
Description
Parathyroid Hormone Protein is fully biologically active when compared to standard. The ED50 as determined by its ability to induce cAMP accumulation in murine MC3T3E1 cells is less than 50 ng/mL, corresponding to a specific activity of >2.0 x 10^4 IU/mg.
Specifications
Specifications
| Accession Number | P01270, P01270, P01270 |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.0. |
| Gene ID (Entrez) | 5741 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 4.1 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/μg of PTH as determined by LAL method. |
| Gene Alias | Parathormone, Parathyrin, parathyroid hormone, parathyroid hormone 1, PTH1 |
| Gene Symbol | PTH |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction