missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human PTEN Protein
Click to view available options
:
10ug; Carrier Free
Description
Measured by its ability to dephosphorylate either diC8PIP3 or diC16PIP3. The specific activity is >480 nmol/min/mg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | P60484 |
| For Use With (Application) | Inhibition Assay |
| Formulation | Supplied as a 0.2 μm filtered solution in HEPES, NaCl, EDTA, Glycerol and Betamercaptoethanol. |
| Gene ID (Entrez) | 5728 |
| Molecular Weight (g/mol) | M.W. (Observed): 59 kDa, reducing conditions; M.W. (theoretical): 48 kDa |
| Name | PTEN |
| Quantity | 10 μg |
| Source | E. coli-derived human PTEN protein Thr2-Val403, with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction