missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PSMG2 Protein

A cDNA sequence encoding the PSMG2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13195-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15942799

  • £5730.00 / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

PSMG2 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMFVPCGE SAPDLAGFTL LMPAVSVGNV GQLAMDLIIS TLNMSKIGYF YTDCLVPMVG NNPYATTEGN STELSINAEV YSLPSRKLVA LQLRSIFIKY KSKPFCEKLL SWVKSSGCAR VIVLSSSHSY QRNDLQLRST PFRYLLTPSM QKSVQNKIKS LNWEEMEKSR CIPEIDDSEF CIRIPGGGIT KTLYDESCSK EIQMAVLLKF VSEGDNIPDA LGLVEYLNEW LQILKPLSDD PTVSASRWKI PSSWRLLFGS GLPPALF
Greater than 90.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
PSMG2 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 40% glycerol and 0.1M NaCl.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers