missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PSMB7 Protein
A cDNA sequence encoding the PSMB7 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13189-1ug
Additional Details : Weight : 0.01000kg
Specifications
PSMB7 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MTTIAGVVYK DGIVLGADTR ATEGMVVADK NCSKIHFISP NIYCCGAGTA ADTDMTTQLI SSNLELHSLS TGRLPRVVTA NRMLKQMLFR YQGYIGAALV LGGVDVTGPH LYSIYPHGST DKLPYVTMGS GSLAAMAVFE DKFRPDMEEE EAKNLVSEAI AAGIFNDLGS GSNIDLCVIS KNKLDFLRPY TVPNKKGTRL GRYRCEKGTT AVLTEKITPL EIEVLEETVQ TMDTS | |
Greater than 80.0% as determined by SDS-PAGE. |
1 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PSMB7 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol and 1mM DTT. |