missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein SSX5 Protein

A DNA sequence encoding the Homo sapiens (Human) Protein SSX5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6734-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16075460

  • £2480.00 / 1mg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_066295.3
Protein SSX5 Protein
Research Use Only
SSX5
Recombinant Protein
E. coli
MNGDDAFVRR PRVGSQIPQK MQKHPWRQVC DRGIHLVNLS PFWKVGREPA SSIKALLCGR GEARAFDDIA KYFSEKEWEK MKASEKIIYV YMKRKYEAMT KLGFKATLPP FMRNKRVADF QGNDFDNDPN RGNQVEHPQM TFGRLQGIFP KITPEKPAEE GNDSKGVPEA SGPQNNGKQL RPSGKLNTSE KVNKTSGPKR GKHAWTHRVR ERKQLVIYEE ISDPQEDDE
Greater than 80% as determined by SDS-PAGE.
6758
1 mg
Not determined.
Full Length of BC016640
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers