missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein prune homolog Protein

Product Code. 16010821
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16010821 100 μg 100µg
16020821 10 μg 10µg
16060821 50 μg 50µg
16040821 200 μg 200µg
16050821 500 μg 500µg
16030821 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Produktkod. 16010821 missing translation for 'mfr' enQuireBio™ missing translation for 'supplierNo' QP7790ec100ug

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

A DNA sequence encoding the Homo sapiens (Human) Protein prune homolog, was expressed in the hosts and tags indicated.

Specifikationer

Accession Number NP_001290158.1
Gene ID (Entrez) 58497
Name Protein prune homolog Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol PRUNE
Protein Length Full Length of Isoform 6
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MLRKDQKTIY RQGVKVAISA IYMDLEICEV LERSHSPPLK LTPASSTHPN LHAYLQGNTQ VSRKKLLPLL QEALSAYFDS MKIPSGQPET ADVSREQVDK ELDRASNSLI SGLSQDEEDP PLPPTPMNSL VDECPLDQGL PKLSAEAVFE KCSQISLSQS TTASLSKK
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.