missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein FAM49B Protein

A DNA sequence encoding the Homo sapiens (Human) Protein FAM49B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8121-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16022781

  • £1735.00 / 1mg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001243692.1
Protein FAM49B Protein
Research Use Only
FAM49B
Recombinant Protein
E. coli
MGNLLKVLTC TDLEQGPNFF LDFENAQPTE SEKEIYNQVN VVLKDAEGIL EDLQSYRGAG HEIREAIQHP ADEKLQEKAW GAVVPLVGKL KKFYEFSQRL EAALRGLLGA LTSTPYSPTQ HLEREQALAK QFAEILHFTL RFDELKMTNP AIQNDFSYYR RTLSRMRINN VPAEGENEVN NELANRMSLF YAEATPMLKT LSDATTKFVS ENKNLPIENT TDCLSTMASV CRVMLETPEY RSRFTNEETV SFCLRVMVGV IILYDHVHPV GAFAKTSKID MKGCIKVLKD QPPNSVEGLL NALRYTTKHL NDETTSKQIK SMLQ
Greater than 90% as determined by SDS-PAGE.
51571
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers