missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein FAM114A2 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein FAM114A2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8153-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001304922.1 | |
Protein FAM114A2 Protein | |
Research Use Only | |
FAM114A2 | |
Recombinant Protein | |
E. coli | |
MSDKDDIETP LLTEAAPILE DGNCEPAKNS ESVDQGAKPE SKSEPVVSTR KRPETKPSSD LETSKVLPIQ DNVSKDVPQT RWGYWGSWGK SILSSASATV ATVGQGISNV IEKAETSLGI PGPSEISTEV KYVAGETNAK ENENSSPVAG AFGVFSTIST AVQSTGKSVI SGGLDALEFI GKKTMDVIAE GDPGFKRTKG LMNRNATLSQ VLREAKEKEE IRTSNEVTVE TDKKTHYGLL FDEFQGLSHL EALEMLSQES EIKVKSILNS LSGEELETLK VELEQLKETF SLAEFCEEEE EEKKGDEDFT KDITELFSQL HVSSKPEKLA RARNTAHEWI RKSLTKPLAE NEEGEKQSEA ENTEQVNKNS IEDIHAFAIR SLAELTACSI ELFHKTAALV LHGRKQEVTA IERSQTLSQM TIVLCKELSS LSKEFTTCLT TAGVKEMADV LNPLITAVFL EASNSASYIQ DAFQLLLPVL EISLIENKIE SHRHELQGQK PLLEH | |
Greater than 90% as determined by SDS-PAGE. |
10827 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |