missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein FAM114A2 Protein

A DNA sequence encoding the Homo sapiens (Human) Protein FAM114A2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8153-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16012941

  • £1735.00 / 1mg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001304922.1
Protein FAM114A2 Protein
Research Use Only
FAM114A2
Recombinant Protein
E. coli
MSDKDDIETP LLTEAAPILE DGNCEPAKNS ESVDQGAKPE SKSEPVVSTR KRPETKPSSD LETSKVLPIQ DNVSKDVPQT RWGYWGSWGK SILSSASATV ATVGQGISNV IEKAETSLGI PGPSEISTEV KYVAGETNAK ENENSSPVAG AFGVFSTIST AVQSTGKSVI SGGLDALEFI GKKTMDVIAE GDPGFKRTKG LMNRNATLSQ VLREAKEKEE IRTSNEVTVE TDKKTHYGLL FDEFQGLSHL EALEMLSQES EIKVKSILNS LSGEELETLK VELEQLKETF SLAEFCEEEE EEKKGDEDFT KDITELFSQL HVSSKPEKLA RARNTAHEWI RKSLTKPLAE NEEGEKQSEA ENTEQVNKNS IEDIHAFAIR SLAELTACSI ELFHKTAALV LHGRKQEVTA IERSQTLSQM TIVLCKELSS LSKEFTTCLT TAGVKEMADV LNPLITAVFL EASNSASYIQ DAFQLLLPVL EISLIENKIE SHRHELQGQK PLLEH
Greater than 90% as determined by SDS-PAGE.
10827
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers