missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein E7 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein E7, was expressed in the hosts and tags indicated.
£154.81 - £1735.00
Specifications
Name | Protein E7 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | E7 |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16028480
|
enQuireBio™
QP7367-EC-100UG |
100 μg |
£403.00
100µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16048480
|
enQuireBio™
QP7367-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16038480
|
enQuireBio™
QP7367-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058480
|
enQuireBio™
QP7367-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16068480
|
enQuireBio™
QP7367-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078480
|
enQuireBio™
QP7367-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Protein E7 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
E7 | |
Recombinant Protein | |
E. coli | |
MHGPKATLQD IVLHLEPQNE IPVDLLCHEQ LSDSEEENDE IDGVNHQHLP ARRAEPQRHT MLCMCCKCEA RIKLVVESSA DDLRAFQQLF LNTLSFVCPW CASQQ | |
Greater than 90% as determined by SDS-PAGE. |