missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Proteasome subunit beta type-4 Protein
A DNA sequence encoding the Homo sapiens (Human) Proteasome subunit beta type-4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8363-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002787.2 | |
Proteasome subunit beta type-4 Protein | |
Research Use Only | |
PSMB4 | |
Recombinant Protein | |
E. coli | |
TQNPMVTGTS VLGVKFEGGV VIAADMLGSY GSLARFRNIS RIMRVNNSTM LGASGDYADF QYLKQVLGQM VIDEELLGDG HSYSPRAIHS WLTRAMYSRR SKMNPLWNTM VIGGYADGES FLGYVDMLGV AYEAPSLATG YGAYLAQPLL REVLEKQPVL SQTEARDLVE RCMRVLYYRD ARSYNRFQIA TVTEKGVEIE GPLSTETNWD IAHMISGFE | |
Greater than 90% as determined by SDS-PAGE. |
5692 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |