missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Proteasome subunit beta type-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Proteasome subunit beta type-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8365-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_002784.1 | |
Proteasome subunit beta type-1 Protein | |
Research Use Only | |
PSMB1 | |
Recombinant Protein | |
E. coli | |
RFSPYVFNGG TILAIAGEDF AIVASDTRLS EGFSIHTRDS PKCYKLTDKT VIGCSGFHGD CLTLTKIIEA RLKMYKHSNN KAMTTGAIAA MLSTILYSRR FFPYYVYNII GGLDEEGKGA VYSFDPVGSY QRDSFKAGGS ASAMLQPLLD NQVGFKNMQN VEHVPLSLDR AMRLVKDVFI SAAERDVYTG DALRICIVTK EGIREETVSL RKD | |
Greater than 90% as determined by SDS-PAGE. |
5689 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |