missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Proteasome subunit beta type-1 Protein

A DNA sequence encoding the Homo sapiens (Human) Proteasome subunit beta type-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8365-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16074241

  • £384.00 / 100µg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_002784.1
Proteasome subunit beta type-1 Protein
Research Use Only
PSMB1
Recombinant Protein
E. coli
RFSPYVFNGG TILAIAGEDF AIVASDTRLS EGFSIHTRDS PKCYKLTDKT VIGCSGFHGD CLTLTKIIEA RLKMYKHSNN KAMTTGAIAA MLSTILYSRR FFPYYVYNII GGLDEEGKGA VYSFDPVGSY QRDSFKAGGS ASAMLQPLLD NQVGFKNMQN VEHVPLSLDR AMRLVKDVFI SAAERDVYTG DALRICIVTK EGIREETVSL RKD
Greater than 90% as determined by SDS-PAGE.
5689
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers