missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Prostasin/Prss8 Protein

Product Code. 18370258
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18370258 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18370258 Supplier R&D Systems™ Supplier No. 4599SE010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to cleave the fluorogenic peptide substrate Boc-QAR-AMC (Catalog # ES014 ). The specific activity is >10 pmol/min/μg, as measured under the described conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_002764
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl and CaCl2.
Gene ID (Entrez) 5652
Molecular Weight (g/mol) M.W. (Observed): 39 kDa, reducing conditions; M.W. (theoretical): 32 kDa (Pro) & 31 kDa (Mature)
Name Prostasin/Prss8
Quantity 10 μg
Source Chinese Hamster Ovary cell line, CHO-derived human Prostasin/Prss8 protein Ala33-Gly319, with a C-terminal 10-His tag The pro form was activated and further purified.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Structural Form Active
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol PRSS8
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein Prostasin/Prss8
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.