missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Proprotein convertase PC4 GST (N-Term) Protein

Product Code. 18333539 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18333539 25 μg 25µg
18380929 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18333539 Supplier Novus Biologicals™ Supplier No. H00054760P0125ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formulation 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 54760
Molecular Weight (g/mol) 52.36 kDa
Quantity 25 μg
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Gene Alias DKFZp434B217, EC 3.4.21, EC 3.4.21.75, EC 3.4.21.93, MGC34749, PC4EC 3.4.21.-, Proprotein convertase 4, proprotein convertase subtilisin/kexin type 4, SPC5
Gene Symbol PCSK4
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein Proprotein convertase PC4
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.