missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Prolactin / PRL Protein

Product Code. 15975969
Click to view available options
Quantity:
1 mg
10 μg
100 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
50µg
This item is not returnable. View return policy

Product Code. 15975969

Brand: enQuireBio™ QP535910ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number NP_000939.1
Gene ID (Entrez) 5617
Name Prolactin / PRL Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol PRL
Biological Activity Shows bioactivity at a concentration of 1 ng/mL in the NB2-11 cell proliferation assay. The bioactivity acceptance criteria is set at 1.0 x 10^6 units/mg.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence MLPICPGGAA RCQVTLRDLF DRAVVLSHYI HNLSSEMFSE FDKRYTHGRG FITKAINSCH TSSLATPEDK EQAQQMNQKD FLSLIVSILR SWNEPLYHLV TEVRGMQEAP EAILSKAVEI EEQTKRLLEG MELIVSQVHP ETKENEIYPV WSGLPSLQMA DEESRLSAYY NLLHCLRRDS HKIDNYLKLL KCRIIHNNNC
Buffer 10 mM sodium phosphate, pH 7.5
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.