missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Proinsulin C-Peptide Analogue Protein
Shop All Bio Techne ProductsSpecifications
Specifications
| Accession Number | P01308, P01308, P01308 |
| For Use With (Application) | Western Blot, PAGE |
| Formulation | Lyophilized from a 0.2 um filtered solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 3630 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 3.6 kDa |
| Quantity | 1000 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/μg of Proinsulin C-Peptide Analogue as determined by LAL method. |
| Gene Symbol | INS |
| Reconstitution | Reconstitute with sterilized distilled water or 0.1% BSA aqueous buffer to a final concentration of 0.1 - 1.0 mg/ml. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?