missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Procollagen C-endopeptidase enhancer 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Procollagen C-endopeptidase enhancer 2, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_037495.1 |
---|---|
Gene ID (Entrez) | 26577 |
Name | Procollagen C-endopeptidase enhancer 2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16083540
|
enQuireBio™
QP6472-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16003550
|
enQuireBio™
QP6472-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16093540
|
enQuireBio™
QP6472-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16013550
|
enQuireBio™
QP6472-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16023550
|
enQuireBio™
QP6472-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16033550
|
enQuireBio™
QP6472-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_037495.1 | |
Procollagen C-endopeptidase enhancer 2 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
26577 | |
Research Use Only | |
PCOLCE2 | |
Recombinant Protein | |
E. coli | |
QSPERPVFTC GGILTGESGF IGSEGFPGVY PPNSKCTWKI TVPEGKVVVL NFRFIDLESD NLCRYDFVDV YNGHANGQRI GRFCGTFRPG ALVSSGNKMM VQMISDANTA GNGFMAMFSA AEPNERGDQY CGGLLDRPSG SFKTPNWPDR DYPAGVTCVW HIVAPKNQLI ELKFEKFDVE RDNYCRYDYV AVFNGGEVND ARRIGKYCGD SPPAPIVSER NELLIQFLSD LSLTADGFIG HYIFRPKKLP TTTEQPVTTT FPVTTGLKPT VALCQQKCRR TGTLEGNYCS SDFVLAGTVI TTITRDGSLH ATVSIINIYK EGNLAIQQAG KNMSARLTVV CKQCPLLRRG LNYIIMGQVG EDGRGKIMPN SFIMMFKTKN QKLLDALKNK QC | |
Greater than 90% as determined by SDS-PAGE. |